missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NQO-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7567-SP
This item is not returnable.
View return policy
Description
NQO-1 Polyclonal specifically detects NQO-1 in Human samples. It is validated for Western Blot.
Specifications
| NQO-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Azoreductase, DHQU, DIA4, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), Diaphorase-4, Dioxin-inducible 1, DTD, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1, NMOR1diaphorase-4, NQO1, Phylloquinone reductase, QR1, QR1NMORI, Quinone reductase 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1728 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| P15559 | |
| NQO1 | |
| E. coli-derived recombinant human NQO-1 Met1-Lys274 Accession # P15559 | |
| 25 μg | |
| Primary | |
| Detects human NQO-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human NQO-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction