missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NPRB/NPR2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9725V-100UG
This item is not returnable.
View return policy
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Flow Cytometry
Specifications
| NPRB/NPR2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK, AMDM, ANPb, ANPRB, ECDM, GC-B, GUC2B, NPR2, NPRBi, SNSK | |
| Mouse myeloma cell line, NS0-derived human NPRB/NPR2, Arg23-Ile458, Accession # P20594 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1060722 | |
| Flow Cytometry | |
| P20594 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4882 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction