Learn More
Human NPRA/NPR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB4860
 View more versions of this product
Product Code. 18122471
  • £380.00 / 100µg
Description
NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPRA/NPR1
Monoclonal
LYOPH
Western Blot 1 ug/mL
P16066
NPR1
Mouse myeloma cell line NS0-derived recombinant human NPRA/NPR1 Gly33-Glu473 Accession # P16066
100 μg
Primary
Detects human NPRA/NPR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse NPR1 or recombinant human NPR2 is observed.
Human
Purified
Western Blot
377029
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4881
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1