missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NPC1 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
NPC1 Synthetic Peptide, PEP-0867, from Invitrogen.
Mutations in the Niemann-Pick disease type C1 (NPC1) gene result in a fatal progressive neurodegenerative disorder characterized by an abnormal sequestration of lipids including cholesterol and glycosphingolipids. The NPC1 protein is a large protein that resides in the limiting membrane of endosomes and lysosomes and mediates intracellular cholesterol trafficking via binding of cholesterol to its N-terminal domain. NPC1 transports low-density lipoproteins to late endosomal/lysosomal compartments where they are hydrolyzed and released as free cholesterol. NPC1, in addition to FTO, MC4R, and PTER has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations.
Specifications
Specifications
| Gene ID (Entrez) | 4864 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O15118 |
| Common Name | Human NPC1 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction