missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human NOV/CCN3 DuoSet ELISA
Description
The Human NOV/CCN3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human NOV/CCN3.
Specifications
Specifications
| Quantity | 15 x 96-well Plates |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4856 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?