missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-1 Intracellular Domain Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3647
This item is not returnable.
View return policy
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Notch-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| AAG33848 | |
| NOTCH1 | |
| E. coli-derived recombinant human Notch-1 Gly2428-Lys2556 Accession # AAG33848 | |
| 100 μg | |
| Primary | |
| Detects human Notch-1 Intracellular Domain in Western blots. In Western blots, this antibody does not cross-react with the corresponding intracellular segments of recombinant human (rh) Notch-2, rhNotch-3, or rhNotch-4. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 433802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4851 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction