missing translation for 'onlineSavingsMsg'
Learn More

Human Notch-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£255.00 - £498.00

Specifications

Antigen Notch-1
Clone 527425
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18759938
GREENER_CHOICE
View Documents
R&D Systems
FAB5317A-025
25 Tests
£255.00
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18796376
GREENER_CHOICE
View Documents
R&D Systems
FAB5317A-100
100 Tests
£498.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Notch-1
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
NOTCH1
IgG1
Protein A or G purified from hybridoma culture supernatant
Detects human Notch-1.
527425
Flow Cytometry
APC
Mouse
Human
P46531
4851
Mouse myeloma cell line NS0-derived recombinant human Notch-1 aa 19-526 Accession # P46531
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.