missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NOSIP (aa 222-299) Control Fragment Recombinant Protein

Product Code. 30213502
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213502

Brand: Invitrogen™ RP105223

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84532 (PA5-84532. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1. 2, Nav1. 3, Nav1. 5, Nav1. 7, Nav1. 8, Kv1. 3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y314
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51070
Name Human NOSIP (aa 222-299) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310061K06Rik; CGI-25; E3 ubiquitin-protein ligase NOSIP; eNOS-interacting protein; nitric oxide synthase interacting protein; nitric oxide synthase-interacting protein; Nosip; RING-type E3 ubiquitin transferase NOSIP
Common Name NOSIP
Gene Symbol NOSIP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VCAVTRDSLSNATPCAVLRPSGAVVTLECVEKLIRKDMVDPVTGDKLTDRDIIVLQRGGTGFAGSGVKLQAEKSRPVM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.