missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NOS1AP (aa 250-354) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56330 (PA5-56330. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Accession Number | O75052 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9722 |
| Name | Human NOS1AP (aa 250-354) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6330408P19Rik; Capon; carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; C-terminal PDZ domain ligand of neuronal nitric oxide synthase; C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Kiaa0464; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; nitric oxide synthase 1 adaptor protein; NOS1AP |
| Common Name | NOS1AP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction