missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Norrin Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3014
This item is not returnable.
View return policy
Description
Norrin/NDP Polyclonal specifically detects Norrin/NDP in Human samples. It is validated for Western Blot.
Specifications
| Norrin/NDP | |
| Polyclonal | |
| Biotin | |
| Q00604 | |
| NDP | |
| E. coli-derived recombinant human Norrin Lys25-Ser133 Accession # Q00604 | |
| 50 μg | |
| Primary | |
| Detects human Norrin in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Norrin is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4693 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction