missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NOP16 (aa 68-135) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57693 (PA5-57693. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NOP16 is transcriptionally regulated by c-Myc (MYC), upregulated in breast cancer, and overexpression is associated with poor patient survival (Butt et al., 2008).
Specifications
Specifications
| Accession Number | Q9Y3C1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51491 |
| Name | Human NOP16 (aa 68-135) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AA409471; CGI-117; D13Wsu177e; HBV pre-S2 trans-regulated protein 3; HSPC111; HSPC185; hypothetical protein HSPC111; Nop16; NOP16 nucleolar protein; NOP16 nucleolar protein homolog; nucleolar protein 16; nucleolar protein 16 homolog; nucleolar protein 16-like; Protein ZH1; RGD1305727 |
| Common Name | NOP16 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction