missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Nogo Receptor/Reticulon-4 Receptor ELISA Kit

Product Code. 17125119
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17125119

Brand: Invitrogen™ EH342RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Nogo Receptor/Reticulon-4 Receptor quantitates human Nogo Receptor/Reticulon-4 Receptor in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nogo Receptor/Reticulon-4 Receptor.

NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number HGNC:1860; NgR; NgR1; nogo 66 receptor; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; reticulon-4 receptor; Rtn4r; UNQ330/PRO526
Assay Range 0.85 to 200 ng/mL
Assay Sensitivity 0.85 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 65078
Gene Symbol RTN4R
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Nogo Receptor
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.