missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NME3 (aa 139-169) Control Fragment Recombinant Protein

Product Code. 30203312
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203312

Brand: Invitrogen™ RP106413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65738 (PA5-65738. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13232
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4832
Name Human NME3 (aa 139-169) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810009F08Rik; AI413736; c371H6.2; DR-nm23; expressed in non-metastatic cells 3; expressed in non-metastatic cells 3 protein (nucleoside diphosphate kinase); expressed in non-metastatic cells 3, protein (nucleoside diphosphate kinase); NDK 3; Ndk3; NDP kinase 3; NDP kinase C; NDPKC; NDPK-C; NM23-dr; NM23H3; nm23-H3; nm23-M3; NME/NM23 nucleoside diphosphate kinase 3; Nme3; non-metastatic cell expressed protein 3; non-metastatic cells 3, protein expressed in; nucleoside diphosphate kinase 3; Nucleoside diphosphate kinase C; nucleoside diphosphate kinase DR-nm23
Common Name NME3
Gene Symbol NME3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SVESARREIALWFRADELLCWEDSAGHWLYE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.