missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NLRP6/NALP6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC9145G-100UG
This item is not returnable.
View return policy
Description
NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| NALP6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NALP6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
| E.coli-derived recombinant human NLRP6/NALP6 Lys35-Arg193 Accession # P59044 | |
| RUO | |
| 171389 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 920631 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P59044 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human NLRP6/NALP6 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction