missing translation for 'onlineSavingsMsg'
Learn More

Human NLRP3/NALP3 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£160.00 - £407.00

Specifications

Antigen NLRP3/NALP3
Clone 447609
Dilution Western Blot 2 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18740377
GREENER_CHOICE
View Documents
R&D Systems
MAB67891-SP
25 μg
£160.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18770137
GREENER_CHOICE
View Documents
R&D Systems
MAB67891-100
100 μg
£407.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NLRP3/NALP3
Western Blot 2 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
AGTAVPRL, AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CIAS1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCAS, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
E. coli-derived recombinant human NALP-3/NALP3, Met3-Val223, Accession # Q96P20
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human NALP-3/NALP3 in direct ELISAs.
447609
Western Blot
Unconjugated
Mouse
Human
Q96P20
114548
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.