missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NLRP2/NALP2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4684
This item is not returnable.
View return policy
Description
NLRP2/NALP2 Polyclonal specifically detects NLRP2/NALP2 in Human samples. It is validated for Western Blot.
Specifications
| NLRP2/NALP2 | |
| Polyclonal | |
| Unconjugated | |
| Q9NX02 | |
| NLRP2 | |
| E. coli-derived recombinant human NLRP2/NALP2 Met1-Thr208 Accession # Q9NX02 | |
| 100 μg | |
| Primary | |
| Detects human NLRP2/NALP2 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CLR19.9, FLJ20510, NACHT, leucine rich repeat and PYD containing 2, NACHT, LRR and PYD domains-containing protein 2, NALP2, NALP2NACHT, LRR and PYD containing protein 2, NBS1, NBS1PYRIN-containing APAF1-like protein 2, NLR family, pyrin domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 2, PAN1, PAN1Nucleotide-binding site protein 1, PYPAF2, PYPAF2PYRIN domain and NACHT domain-containing protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55655 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction