missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NLRP2/NALP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4684-SP
This item is not returnable.
View return policy
Description
NLRP2/NALP2 Monoclonal specifically detects NLRP2/NALP2 in Human samples. It is validated for Western Blot.
Specifications
| NLRP2/NALP2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NX02 | |
| NLRP2 | |
| E. coli-derived recombinant human NLRP2/NALP2 Met1-Thr208 Accession # Q9NX02 | |
| 25 μg | |
| Primary | |
| Detects human NLRP2/NALP2 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 443625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLR19.9, FLJ20510, NACHT, leucine rich repeat and PYD containing 2, NACHT, LRR and PYD domains-containing protein 2, NALP2, NALP2NACHT, LRR and PYD containing protein 2, NBS1, NBS1PYRIN-containing APAF1-like protein 2, NLR family, pyrin domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 2, PAN1, PAN1Nucleotide-binding site protein 1, PYPAF2, PYPAF2PYRIN domain and NACHT domain-containing protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55655 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction