missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NKX2-8 Partial ORF (NP_055175.2, 3 a.a. - 50 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026257-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075385

  • £429.00 / 25µg
Estimated Shipment: 09-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: TSGRLSFTVRSLLDLPEQDAQHLPRREPEPRAPQPDPCAAWLDSERGH
Specifications

Specifications

NP_055175.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.02kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSGRLSFTVRSLLDLPEQDAQHLPRREPEPRAPQPDPCAAWLDSERGH
RUO
NKX2-8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26257
NKX2-8 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NKX2.8/NKX2H/Nkx2-9
NKX2-8
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers