missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKX2.5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2444
This item is not returnable.
View return policy
Description
NKX2.5 Monoclonal specifically detects NKX2.5 in Human samples. It is validated for Immunohistochemistry.
Specifications
| NKX2.5 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P52952 | |
| NKX2-5 | |
| E. coli-derived recombinant human NKX2.5 Gln24-Arg137 Accession # P52952 | |
| 100 μg | |
| Primary | |
| Detects human NKX-2.5 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 259416 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2-5, NKX2E, NKX2EFLJ97197, NKX4-1, tinman paralog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1482 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction