missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Nkx2.3 (aa 62-146) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61547 (PA5-61547. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NKX2-3 encodes a homeodomain-containing transcription factor. The encoded protein is a member of the NKX family of homeodomain transcription factors. Studies of similar proteins in mouse and rat have indicated a potential role in cellular differentiation.
Spécification
Spécification
| Accession Number | Q8TAU0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 159296 |
| Name | Human Nkx2.3 (aa 62-146) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | CS x 3; Drosophila NK2 transcription factor related, locus 3; homeobox protein NK-2 homolog 3; Homeobox protein NK-2 homolog C; homeobox protein Nkx-2.3; homeobox protein Nkx-2.3; LOW QUALITY PROTEIN: homeobox protein Nkx-2.3; hypothetical protein LOC539451; NK2 homeobox 3; NK2 transcription factor related, locus 3; NK2.3; NK x 2.3; Nkx-2.3; NK x 23; NK x 2-3; NK x 2 C; Nk x 2-C; NK x 4-3; tinman |
| Common Name | Nkx2.3 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu