missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp80/KLRF1 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£194.00 - £409.00
Specifications
| Antigen | NKp80/KLRF1 |
|---|---|
| Clone | 239127 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736177
![]() |
R&D Systems
FAB1900P-025 |
25 Tests |
£194.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797876
![]() |
R&D Systems
FAB1900P-100 |
100 Tests |
£409.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp80/KLRF1 Monoclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry.Specifications
| NKp80/KLRF1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| KLRF1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NKp80/KLRF1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or recombinant mouse NKp46 is observed. |
| 239127 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Human | |
| Q9NZS2 | |
| 51348 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 Val66-Tyr231 Accession # Q9NZS2 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title