missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp80/KLRF1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1900V-100UG
This item is not returnable.
View return policy
Description
NKp80/KLRF1 Monoclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKp80/KLRF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9NZS2 | |
| KLRF1 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1 Val66-Tyr231 Accession # Q9NZS2 | |
| RUO | |
| 51348 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 239127 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Activating coreceptor NKp80, CLEC5C, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, KLRF1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human NKp80/KLRF1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp44, rhNKp46, or recombinant mouse NKp46 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction