missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp46/NCR1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB1850A-100
This item is not returnable.
View return policy
Description
NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKp46/NCR1 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| NCR1 | |
| Mouse T cell hybridoma transfected with human NKp46/NCR1 | |
| 100 Tests | |
| Primary | |
| Detects human NKp46/NCR1 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 195314 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9437 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction