missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp46/NCR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 22 publications
£189.00 - £1895.00
Specifications
| Antigen | NKp46/NCR1 |
|---|---|
| Clone | 195314 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Agonist Activity, Immunocytochemistry 8-25 ug/mL, CyTOF-reported |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, In vitro Assay, Immunocytochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780048
![]() |
R&D Systems
MAB1850-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18160801
![]() |
R&D Systems
MAB1850-100 |
100 μg |
£481.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702817
![]() |
R&D Systems
MAB1850-500 |
500 μg |
£1895.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF-reported.Specifications
| NKp46/NCR1 | |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunohistochemistry, In vitro Assay, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 9437 | |
| Mouse T cell hybridoma transfected with human NKp46/NCR1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human NKp46/NCR1 in direct ELISAs and Western blots. |
| 195314 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Agonist Activity, Immunocytochemistry 8-25 ug/mL, CyTOF-reported | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| NCR1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title