missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp30/NCR3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£176.00 - £469.00
Specifications
| Antigen | NKp30/NCR3 |
|---|---|
| Clone | 210845 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776357
![]() |
R&D Systems
FAB1849A-025 |
25 Tests |
£176.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705919
![]() |
R&D Systems
FAB1849A-100 |
100 Tests |
£469.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry.Specifications
| NKp30/NCR3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 259197 | |
| Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 210845 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS | |
| NCR3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NKp30/NCR3 on cell transfectants and peripheral blood NK cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title