missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NKp30 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH339RBX10
Additional Details : Weight : 2.50000kg
Description
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].Specifications
O14931 | |
0.085 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Ncr3 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.085 to 20 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
259197 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL|Serum, 50 μL|Plasma, 50 μL | |
NKp30 | |
4 hr 45 min |