missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NKp30 ELISA Kit
Human NKp30 quantitates human NKp30 in serum, plasma, supernatant.
£540.00 - £2320.00
Specifications
Accession Number | 1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; Natural killer cell p30-related protein; natural killer cell receptor; NCR3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED |
---|---|
Assay Range | 0.085 to 20 ng/mL |
Assay Sensitivity | 0.085 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17185109
|
Invitrogen™
EH339RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17184633
|
Invitrogen™
EH339RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Human NKp30 quantitates human NKp30 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NKp30.
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].Specifications
1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; Natural killer cell p30-related protein; natural killer cell receptor; NCR3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED | |
0.085 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Ncr3 | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
NKp30 | |
4 hr 45 min |
0.085 to 20 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O14931 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |