missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2D/CD314 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1392A-100
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry
Specifications
| NKG2D/CD314 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing Tris and Sodium Azide | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| Mouse myeloma cell line NS0-derived recombinant human NKG2D/CD314, Phe78-Val216, Accession # P26718 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1046002 | |
| 10 μL/106 cells | |
| P26718 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22914 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction