missing translation for 'onlineSavingsMsg'
Learn More

Human NKG2C/CD159c Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 11 publications

£170.00 - £442.00

Specifications

Antigen NKG2C/CD159c/KLRC2
Clone 134522
Concentration LYOPH
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported
Applications Flow Cytometry, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18754319
GREENER_CHOICE
View Documents
R&D Systems
MAB1381-SP
25 μg
£170.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18720068
GREENER_CHOICE
View Documents
R&D Systems
MAB1381-100
100 μg
£442.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.
Specifications

Specifications

NKG2C/CD159c/KLRC2
LYOPH
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor
KLRC2
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
134522
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
3822
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human NKG2C/CD159c. Detects human NKG2C/CD159c as part of the NKG2C/CD94 heterodimer in flow cytometry. No cross-reactivity with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer is detected.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.