Learn More
Human NKG2A/CD159a PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 26 publications
Brand:  R&D Systems FAB1059P-100
 View more versions of this product
Product Code. 18746286
  • £554.00 / 100 tests
Description
NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKG2A/CD159a/KLRC1
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
KLRC1
BaF3 mouse pro-B cell line transfected with human NKG2A and CD94
100 Tests
Primary
Recognizes the human NKG2A/CD94 heterodimer. It does not recognize the NKG2C/CD94 heterodimer or the CD94 homodimer.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a
Flow Cytometry
131411
Flow Cytometry 10 uL/10^6 cells
CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3821
Human
Purified