Learn More
Human NKG2A/CD159a Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 12 publications
Brand:  R&D Systems MAB1059-100
 View more versions of this product
Product Code. 18773798
  • £467.00 / 100µg
Description
NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKG2A/CD159a/KLRC1
Monoclonal
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3821
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Flow Cytometry, CyTOF
131411
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
KLRC1
BaF3 mouse pro-B cell line transfected with human NKG2A and CD94
100 μg
Primary
Recognizes the human NKG2A/CD94 heterodimer. It does not recognize the NKG2C/CD94 heterodimer or the CD94 homodimer.
Human
Supernatant