missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2A/CD159a Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB1059N-100
This item is not returnable.
View return policy
Description
NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKG2A/CD159a/KLRC1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| KLRC1 | |
| BaF3 mouse pro-B cell line transfected with human NKG2A and CD94 | |
| 100 Tests | |
| Primary | |
| Recognizes the human NKG2A/CD94 heterodimer. It does not recognize the NKG2C/CD94 heterodimer or the CD94 homodimer. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 131411 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3821 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction