missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NK1R Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB66871
This item is not returnable.
View return policy
Description
NK1R Monoclonal specifically detects NK1R in Human samples. It is validated for Immunocytochemistry.
Specifications
| NK1R | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P25103 | |
| TACR1 | |
| HEK293 human embryonic kidney cell line transfected with human NK1R Accession # P25103 | |
| 100 μg | |
| Primary | |
| Detects human NK1R in ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 694501 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPR, SPRsubstance-P receptor, TAC1R, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor, TACR1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6869 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction