missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NIPA4 (aa 345-404) Control Fragment Recombinant Protein

Product Code. 30211783
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211783

Brand: Invitrogen™ RP96939

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58315 (PA5-58315. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NIPAL4 acts as a Mg(2+) transporter and can also transport other divalent cations such as Ba(2+), Mn(2+), Sr(2+) and Co(2+) but to a much less extent than Mg(2+). NIPAL4 may also be a receptor for ligands (trioxilins A3 and B3) from the hepoxilin pathway. The NIPAL4 gene likely encodes a membrane receptor. Mutations in the NIPAL4 gene have been associated with autosomal recessive congenital ichthyosis. Diseases associated with NIPAL4 include Ichthyosis, Congenital, Autosomal Recessive 6 and Autosomal Recessive Congenital Ichthyosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q0D2K0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 348938
Name Human NIPA4 (aa 345-404) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9530066K23Rik; ARCI6; ICHN; Ichthyin; ICHYN; Magnesium transporter NIPA4; NIPA like domain containing 4; Nipa4; NIPAL4; NIPA-like domain containing 4; NIPA-like protein 4; Non-imprinted in Prader-Willi/Angelman syndrome region protein 4; non-imprinted in Prader-Willi/Angelman syndrome region protein 4 homolog; RGD1309452
Common Name NIPA4
Gene Symbol NIPAL4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FKDLDISCASLPHMHKNPPPSPAPEPTVIRLEDKNVLVDNIELASTSSPEEKPKVFIIHS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.