missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NIPA (aa 5-108) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82664 (PA5-82664. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an F-box-containing protein that is a component of an SCF-type E3 ubiquitin ligase complex that regulates the onset of cell division. The G2/M transition in the cell cycle requires the interaction of the proteins cyclin B1 and cyclin-dependent kinase 1. The activated ubiquitin ligase complex targets the protein cyclin B1 for degradation, preventing this transition to mitosis.
Specifications
Specifications
| Accession Number | Q86WB0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51530 |
| Name | Human NIPA (aa 5-108) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110054L24Rik; AU018540; AU019789; C3HC type 1; hematopoietic stem/progenitor cell protein 216; hNIPA; HSPC216; mNIPA; Nipa; nuclear interacting partner of ALK; nuclear interacting partner of anaplastic lymphoma kinase (Alk); nuclear-interacting partner of ALK; Nuclear-interacting partner of anaplastic lymphoma kinase; OTTHUMP00000212330; RGD1561099; ZC3HC1; zinc finger C3HC-type containing 1; zinc finger C3HC-type protein 1; zinc finger, C3HC type 1; zinc finger, C3HC-type 1; zinc finger, C3HC-type containing 1 |
| Common Name | NIPA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction