missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ninjurin-1 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF5105
This item is not returnable.
View return policy
Description
Ninjurin-1 Polyclonal specifically detects Ninjurin-1 in Human samples. It is validated for Western Blot.
Specifications
| Ninjurin-1 | |
| Polyclonal | |
| Biotin | |
| Q92982 | |
| NINJ1 | |
| E. coli-derived recombinant human Ninjurin-1 Asp2-Val81 Accession # Q92982 | |
| 50 μg | |
| Primary | |
| Detects human Ninjurin-1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Ninjurin-2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJ1, NINJURIN, ninjurin 1, ninjurin-1, Ninjurin1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4814 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction