missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nidogen-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3385
This item is not returnable.
View return policy
Description
Nidogen-2 Polyclonal specifically detects Nidogen-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Nidogen-2 | |
| Polyclonal | |
| Unconjugated | |
| Q14112 | |
| NID2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Nidogen-2 Leu31-Lys1375 (Gly832Ala) Accession # Q14112 | |
| 100 μg | |
| Primary | |
| Detects human Nidogen-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human Nidogen-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| NID2, NID-2, nidogen 2 (osteonidogen), nidogen-2, Nidogen2, osteonidogen | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 22795 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction