missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGL-3/LRRC4B Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6919
This item is not returnable.
View return policy
Description
NGL-3/LRRC4B/LRCH4B Monoclonal specifically detects NGL-3/LRRC4B/LRCH4B in Human samples. It is validated for Immunohistochemistry.
Specifications
| NGL-3/LRRC4B/LRCH4B | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9NT99 | |
| LRRC4B | |
| Chinese hamster ovary cell line CHO-derived recombinant human NGL-3/LRRC4B Ala47-Lys576 Accession # Q9NT99 | |
| 100 μg | |
| Primary | |
| Detects human NGL-3/LRRC4B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LRRC3, 4, or 32 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 702021 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp761A179, HSM, Leucine Rich Repeat Containing 4B, Leucine-Rich Repeat-Containing Protein 4B, Leucine-Rich Repeats And Immunoglobulin-Like Domains 4, LRIG4, LRRC4B, Netrin-G3 Ligand, NGL3, NGL-3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 94030 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction