missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGL-1/LRRC4C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4899-SP
This item is not returnable.
View return policy
Description
NGL-1/LRRC4C Polyclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Western Blot.
Specifications
| NGL-1/LRRC4C | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C, LRRC4C, NGL1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57689 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9HCJ2 | |
| LRRC4C | |
| Mouse myeloma cell line NS0-derived recombinant human NGL-1/LRRC4C Gln45-Lys527, predicted Accession # Q9HCJ2 | |
| 25 μg | |
| Primary | |
| Detects human NGL-1/LRRC4C in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction