missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGFR/TNFRSF16 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB367A
This item is not returnable.
View return policy
Description
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Flow Cytometry.
Specifications
| NGFR/TNFRSF16/p75NTR | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4804 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 74902 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P08138 | |
| NGFR | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NGF R Lys29-Asn250 Accession # P08138 | |
| 100 Tests | |
| Primary | |
| Detects human NGF R in direct ELISAs and Western blots. In direct ELISAs, no crossreactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD40, rhBAFF R, rhCD30, rhDR3, rhDR6, rhEDAR, rhFas, rhHVEM, rhGITR, rhLTR B, recominant mouse (rm) NGF R, rhOPG, rmOX40, rhRANK, rhTAJ, rhTNF RI or rhTNF RII is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction