missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGFR/TNFRSF16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £429.00
Specifications
| Antigen | NGFR/TNFRSF16/p75NTR |
|---|---|
| Clone | 1029664 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743617
![]() |
R&D Systems
MAB3671-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702047
![]() |
R&D Systems
MAB3671-100 |
100 μg |
£429.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Immunohistochemistry.Specifications
| NGFR/TNFRSF16/p75NTR | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human NGFR/TNFRSF16 protein, Lys29-Asn250, Accession # P08138 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1029664 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08138 | |
| 4804 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title