missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGFI-B gamma/NOR-1/NR4A3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems PP-H7833-00
This item is not returnable.
View return policy
Description
NGFI-B gamma/NOR-1/NR4A3 Monoclonal specifically detects NGFI-B gamma/NOR-1/NR4A3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NGFI-B gamma/NOR-1/NR4A3 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8013 | |
| Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| H7833 | |
| Western Blot, Immunoprecipitation | |
| NR4A3 | |
| Recombinant human NGFI-Bγ (amino acids 2 - 99) and mouse myeloma cells (NS-1) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction