missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NG-2 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH337RBX10
Additional Details : Weight : 2.50000kg
Description
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.Specifications
Q6UVK1 | |
1.23 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Cspg4 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
1.23 to 300 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
1464 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL|Serum, 50 μL|Plasma, 50 μL | |
NG2 | |
4 hr 45 min |