missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NG-2 ELISA Kit
Human NG-2 quantitates human NG-2 in serum, plasma, supernatant.
£540.00 - £2320.00
Specifications
Accession Number | 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; Melanoma chondroitin sulfate proteoglycan; melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; NG2; proteoglycan AN2 |
---|---|
Assay Range | 1.23 to 300 ng/mL |
Assay Sensitivity | 1.23 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17165109
|
Invitrogen™
EH337RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17164633
|
Invitrogen™
EH337RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Human NG-2 quantitates human NG-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NG-2.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.Specifications
4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; Melanoma chondroitin sulfate proteoglycan; melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; NG2; proteoglycan AN2 | |
1.23 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Cspg4 | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
NG2 | |
4 hr 45 min |
1.23 to 300 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q6UVK1 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |