missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NFE2L1 (aa 680-752) Control Fragment Recombinant Protein

Product Code. 30201060
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201060

Brand: Invitrogen™ RP103600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64222 (PA5-64222. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for 'nuclear respiratory factor 1' which has an official symbol of NRF1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14494
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4779
Name Human NFE2L1 (aa 680-752) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA408798; AW212678; bZIP protein; endoplasmic reticulum membrane sensor NFE2L1; HBZ17; Lcrf1; LCR-F1; locus control region-factor 1; Nfe2l1; NF-E2-related factor 1; NFE2-related factor 1; Nrf1; nuclear factor (erythroid-derived 2)-like 1; nuclear factor erythroid 2-like 1; nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2 like 1; nuclear factor, erythroid 2-like 1; nuclear factor, erythroid derived 2, like 1; nuclear factor, erythroid derived 2,-like 1; Protein NRF1, p110 form; Protein NRF1, p120 form; TCF11; TCF-11; transc; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1; Transcription factor NRF1
Common Name NFE2L1
Gene Symbol NFE2L1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LNLERDVEDLQRDKARLLREKVEFLRSLRQMKQKVQSLYQEVFGRLRDENGRPYSPSQYALQYAGDGSVLLIP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.