missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NFATC1 (aa 88-167) Control Fragment Recombinant Protein

Product Code. 30197606
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197606

Brand: Invitrogen™ RP105543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111680 (PA5-111680. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95644
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4772
Name Human NFATC1 (aa 88-167) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2210017P03Rik; AI449492; AV076380; MGC138448; NFAT tra; NFAT trans; NFAT transcription complex cytosolic component; NFAT2; NFATc; NF-ATC; NFATC1; NF-ATc1; NF-ATc1.2; Nfatcb; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 1; nuclear factor of activated T-cells 1; nuclear factor of activated T-cells'c'; nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor NF-ATc
Common Name NFATC1
Gene Symbol NFATC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GYGAALDGGPAGYFLSSGHTRPDGAPALESPRIEITSCLGLYHNNNQFFHDVEVEDVLPSSKRSPSTATLSLPSLEAYRD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.