missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF2/Merlin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5616-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NF2/Merlin Monoclonal specifically detects NF2/Merlin in Human samples. It is validated for Western Blot.
Specifications
| NF2/Merlin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P35240 | |
| NF2 | |
| E. coli-derived recombinant human NF2/Merlin Met363-Lys578 Accession # P35240 | |
| 25 μg | |
| Primary | |
| Detects human NF2/Merlin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human NF1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 534428 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACN, BANF, Merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4771 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction