missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Neutrophil Elastase/ELA2 ELISA Kit (Colorimetric)

Product Code. 18778473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778473 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18778473 Supplier Novus Biologicals™ Supplier No. NBP191266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human Neutrophil Elastase/ELA2 ELISA Kit (Colorimetric) measures human Neutrophil Elastase/ELA2 in plasma.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 - 10.0ng/mL
Assay Sensitivity 1.98pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1991
Gene Symbol ELANE
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human PMN Elastase, HRP-Conjugate anti-alpha1-PI polyclonal antibody, ready to use, Human PMN Elastase Standard lyophilized, 10ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Wash Buffer Concentrate (10x), Substrate Solution (tetramethyl-benzidine), Stop Solution (2M hydrochloric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Cancer, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil el
Target Neutrophil Elastase/ELA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.