Learn More
Gibco™ Human Neurturin Recombinant Protein, PeproTech®
Description
450-11-1MG will be provided as 2 x 500 μg (450-11-500UG). The functional form of human neurturin is a disulfide-linked homodimer, of two 11.8 kDa polypeptide monomers (204 total amino acid residues). Each monomer contains seven conserved cysteine residues, one of which (Cys 69) is used for inter-chain disulfide bridging, and the others are involved in the intramolecular ring formation known as the cysteine-knot configuration. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q99748 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 4902 |
| Molecular Weight (g/mol) | 23.6 kDa |
| Name | Human Neurturin |
| Quantity | 20 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.