missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurturin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB387
This item is not returnable.
View return policy
Description
Neurturin Monoclonal specifically detects Neurturin in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| Neurturin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| neurturin, NRTN, NTN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4902 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 126422 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q99748 | |
| NRTN | |
| E. coli-derived recombinant human Neurturin Ala96-Val197 Accession # Q99748 | |
| 500 μg | |
| Primary | |
| Detects human Neurturin in direct ELISAs and Western blots. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse Neurturin is observed and no cross-reactivity with recombinant mouse (rm) Artemin, recombinant human (rh) Cripto-1, recombinant Drosophila DPP, rhGDNF, rhLefty A, rmLefty-1, rhMIS, rmNodal, rmPersephin, rhTGF-beta 1, rhLatent TGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2 or rhTGF-beta 3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction